Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC9 SIRNA Assays ›
          Gene Symbol:
          HDAC9
          Gene Name:
          histone deacetylase 9
          Gene Aliases:
          HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
          Chromosome Location:
          Chr.7: 18086942 - 19003745 on Build GRCh38
          UniGene ID:
          Hs.196054
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18774

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204144.2 NP_001191073.1 10, 11 1390
          NM_001204145.2 NP_001191074.1 8, 9 1082
          NM_001204146.2 NP_001191075.1 8, 9 1260
          NM_001204147.2 NP_001191076.1 7, 8 1042
          NM_001204148.2 NP_001191077.1 8, 9 1189
          NM_001321868.1 NP_001308797.1 9, 10 1330
          NM_001321869.1 NP_001308798.1 10, 11 1471
          NM_001321870.1 NP_001308799.1 10, 11 1462
          NM_001321871.1 NP_001308800.1 9, 10 1345
          NM_001321872.1 NP_001308801.1 9, 10 1339
          NM_001321873.1 NP_001308802.1 9, 10 1333
          NM_001321874.1 NP_001308803.1 9, 10 1392
          NM_001321875.1 NP_001308804.1 9, 10 1398
          NM_001321876.1 NP_001308805.1 9, 10 1386
          NM_001321877.1 NP_001308806.1 8, 9 1260
          NM_001321878.1 NP_001308807.1 9, 10 1383
          NM_001321879.1 NP_001308808.1 8, 9 1254
          NM_001321884.1 NP_001308813.1 9, 10 1386
          NM_001321885.1 NP_001308814.1 8, 9 1275
          NM_001321886.1 NP_001308815.1 10, 11 1442
          NM_001321887.1 NP_001308816.1 10, 11 1276
          NM_001321888.1 NP_001308817.1 8, 9 1076
          NM_001321889.1 NP_001308818.1 9, 10 1313
          NM_001321890.1 NP_001308819.1 9, 10 1377
          NM_001321891.1 NP_001308820.1 8, 9 1251
          NM_001321893.1 NP_001308822.1 9, 10 1148
          NM_001321894.1 NP_001308823.1 9, 10 1157
          NM_001321895.1 NP_001308824.1 8, 9 1091
          NM_001321896.1 NP_001308825.1 10, 11 1561
          NM_001321897.1 NP_001308826.1 9, 10 1157
          NM_001321898.1 NP_001308827.1 8, 9 1106
          NM_001321899.1 NP_001308828.1 10, 11 1289
          NM_001321900.1 NP_001308829.1 9, 10 1383
          NM_001321901.1 NP_001308830.1 9, 10 1217
          NM_001321902.1 NP_001308831.1 9, 10 1223
          NM_014707.3 NP_055522.1 9, 10 1214
          NM_058176.2 NP_478056.1 8, 9 1069
          NM_178423.2 NP_848510.1 9, 10 1214
          NM_178425.3 NP_848512.1 8, 9 1078
          XM_011515629.2 XP_011513931.1 9, 10 1172
          XM_011515631.2 XP_011513933.1 10, 11 1486
          XM_011515634.2 XP_011513936.1 8, 9 1040
          XM_011515635.2 XP_011513937.1 10, 11 2038
          XM_011515636.2 XP_011513938.1 9, 10 1759
          XM_011515637.2 XP_011513939.1 9, 10 1183
          XM_011515638.2 XP_011513940.1 9, 10 1753
          XM_011515639.2 XP_011513941.1 9, 10 1950
          XM_011515641.2 XP_011513943.1 10, 11 2035
          XM_011515642.2 XP_011513944.1 8, 9 2764
          XM_011515644.2 XP_011513946.1 10, 11 2010
          XM_011515645.2 XP_011513947.1 8, 9 2749
          XM_011515646.2 XP_011513948.1 8, 9 2740
          XM_011515648.2 XP_011513950.1 9, 10 1768
          XM_011515649.2 XP_011513951.1 9, 10 1972
          XM_011515651.1 XP_011513953.1 8, 9 1006
          XM_017012826.1 XP_016868315.1 9, 10 1744
          XM_017012827.1 XP_016868316.1 10, 11 1827
          XM_017012828.1 XP_016868317.1 8, 9 1820
          XM_017012829.1 XP_016868318.1 9, 10 1147
          XM_017012830.1 XP_016868319.1 9, 10 1138
          XM_017012831.1 XP_016868320.1 8, 9 1015
          XM_017012832.1 XP_016868321.1 9, 10 1147
          GenBank mRNA AB018287.1 - 9, 10 1178
          AF124924.1 - 6, 7 736
          AJ459808.1 - 9, 10 1178
          AK097178.1 - 8, 9 1040
          AK296214.1 - 8, 9 992
          AK297404.1 - 8, 9 1036
          AK304298.1 - 10, 11 1383
          AK304343.1 - 8, 9 1109
          AK304410.1 - 7, 8 1005
          AK316109.1 - 8, 9 1211
          AY032737.1 - 8, 9 1028
          AY032738.1 - 8, 9 1028
          AY197371.1 - 8, 9 1037
          BC111735.1 - 7, 8 904
          BC150328.1 - 9, 10 1189 8860495
          BC152405.1 - 9, 10 1178 8862508
          CR749550.1 - 8, 9 1145

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          transcription, DNA-templated
          inflammatory response
          heart development
          chromatin modification
          histone deacetylation
          neuron differentiation
          B cell differentiation
          cellular response to insulin stimulus
          peptidyl-lysine deacetylation
          B cell activation
          negative regulation of transcription, DNA-templated
          regulation of skeletal muscle fiber development
          regulation of striated muscle cell differentiation
          histone H3 deacetylation
          histone H4 deacetylation
          positive regulation of cell migration involved in sprouting angiogenesis
          transcription corepressor activity
          histone deacetylase activity
          protein kinase C binding
          protein binding
          transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          histone deacetylase binding
          metal ion binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0