Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BARD1 SIRNA Assays ›
          Gene Symbol:
          BARD1
          Gene Name:
          BRCA1 associated RING domain 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 214725645 - 214809711 on Build GRCh38
          UniGene ID:
          Hs.591642
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1886

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BARD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000465.3 NP_000456.2 8 1915
          NM_001282543.1 NP_001269472.1 7 1858
          NM_001282545.1 NP_001269474.1 4 562
          NM_001282548.1 NP_001269477.1 3 505
          NR_104212.1 7 1766
          NR_104215.1 6 1709
          NR_104216.1 7 965
          XM_017004613.1 XP_016860102.1 9 1963
          XM_017004614.1 XP_016860103.1 9 1963
          GenBank mRNA AK223409.1 - 8 1909
          AK301843.1 - 7 1728
          AK314260.1 - 8 1875
          AX794378.1 - 8 1846
          BC126426.1 - 8 1843 8992142
          BC126428.1 - 8 1843 8992144
          BX098964.1 - 3 297
          JF790280.1 - 7 1789
          JF790281.1 - 6 1640
          JF790282.1 - 7 896
          JF790283.1 - 4 448
          JF790284.1 - 3 436
          U76638.1 - 8 1846

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          tissue homeostasis
          DNA replication
          double-strand break repair via nonhomologous end joining
          cellular response to DNA damage stimulus
          cell cycle arrest
          protein ubiquitination
          negative regulation of mRNA 3'-end processing
          regulation of phosphorylation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of protein catabolic process
          negative regulation of protein export from nucleus
          protein K6-linked ubiquitination
          regulation of signal transduction by p53 class mediator
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          kinase binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0