Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BARD1 SIRNA Assays ›
          Gene Symbol:
          BARD1
          Gene Name:
          BRCA1 associated RING domain 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 214725645 - 214809711 on Build GRCh38
          UniGene ID:
          Hs.591642
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1887

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BARD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000465.3 NP_000456.2 7 1772
          NM_001282543.1 NP_001269472.1 6 1715
          NM_001282545.1 NP_001269474.1 3 419
          NM_001282548.1 NP_001269477.1 2 362
          NR_104212.1 6 1623
          NR_104215.1 5 1566
          NR_104216.1 6 822
          XM_017004613.1 XP_016860102.1 8 1820
          XM_017004614.1 XP_016860103.1 8 1820
          GenBank mRNA AK223409.1 - 7 1766
          AK301843.1 - 6 1585
          AK314260.1 - 7 1732
          AX794378.1 - 7 1703
          BC126426.1 - 7 1700 8992142
          BC126428.1 - 7 1700 8992144
          BX098964.1 - 2 154
          JF790280.1 - 6 1646
          JF790281.1 - 5 1497
          JF790282.1 - 6 753
          JF790283.1 - 3 305
          JF790284.1 - 2 293
          U76638.1 - 7 1703

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          tissue homeostasis
          DNA replication
          double-strand break repair via nonhomologous end joining
          cellular response to DNA damage stimulus
          cell cycle arrest
          protein ubiquitination
          negative regulation of mRNA 3'-end processing
          regulation of phosphorylation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of protein catabolic process
          negative regulation of protein export from nucleus
          protein K6-linked ubiquitination
          regulation of signal transduction by p53 class mediator
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          kinase binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0