Search Thermo Fisher Scientific
Gene Symbol: Klrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085405.1 | NP_001078874.1 | 3 | 239 | ||
XM_008763334.1 | XP_008761556.1 | 3 | 297 | |||
XM_008763335.1 | XP_008761557.1 | 3 | 297 | |||
GenBank mRNA | DQ113420.1 | - | 3 | 239 | ||
DQ157011.1 | - | 3 | 239 | |||
DQ157012.1 | - | 3 | 239 | |||
EF100680.1 | - | 3 | 239 |