Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIF14 SIRNA Assays ›
          Gene Symbol:
          KIF14
          Gene Name:
          kinesin family member 14
          Gene Aliases:
          MKS12
          Chromosome Location:
          Chr.1: 200551497 - 200620791 on Build GRCh38
          UniGene ID:
          Hs.3104
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19263

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIF14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001305792.1 NP_001292721.1 9 2124
          NM_014875.2 NP_055690.1 11 2536
          XM_011510231.2 XP_011508533.1 12 2630
          XM_011510232.2 XP_011508534.1 11 2292
          XM_011510233.2 XP_011508535.1 12 2423
          XM_011510235.2 XP_011508537.1 12 2135
          XM_011510236.1 XP_011508538.1 10 2419
          XM_017003005.1 XP_016858494.1 10 2713
          XM_017003006.1 XP_016858495.1 11 2321
          XM_017003007.1 XP_016858496.1 11 1848
          GenBank mRNA BC098582.1 - 1 129
          BC113742.1 - 10 2175 8322794
          D26361.1 - 11 2536

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          microtubule-based movement
          microtubule depolymerization
          mitotic metaphase plate congression
          positive regulation of cell proliferation
          regulation of G2/M transition of mitotic cell cycle
          cerebellar granular layer structural organization
          cerebellar Purkinje cell layer structural organization
          cerebellar cortex development
          hippocampus development
          olfactory bulb development
          cell proliferation in forebrain
          cerebral cortex development
          regulation of cell adhesion
          regulation of cell migration
          cytoskeleton-dependent intracellular transport
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          regulation of myelination
          activation of protein kinase activity
          positive regulation of cytokinesis
          regulation of Rap protein signal transduction
          negative regulation of integrin activation
          substrate adhesion-dependent cell spreading
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          establishment of protein localization
          cell division
          response to docetaxel trihydrate
          mitotic cell cycle process
          regulation of cell maturation
          regulation of G1/S transition of mitotic cell cycle
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP-dependent microtubule motor activity, plus-end-directed
          tubulin binding
          ATPase activity
          protein kinase binding
          PDZ domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline