Search
Search

CRYZL1| Gene Symbol: CRYZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145858.2 | NP_665857.2 | 8, 9 | 760 | ||
| GenBank mRNA | AF029689.1 | - | 8, 9 | 627 | ||
| AF098798.1 | - | 8, 9 | 627 | |||
| AK001293.1 | - | 8, 9 | 653 | |||
| AK315707.1 | - | 8, 9 | 620 | |||
| BC033023.2 | - | 8, 9 | 614 | 5269248 | ||
| BI599245.1 | - | 8, 9 | 663 | |||
| BX648547.1 | - | 9, 10 | 774 | |||