Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Vom2r22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099656.1 | NP_001093126.1 | 1727 | |||
XM_003750425.1 | XP_003750473.1 | 6 | 1727 |
Gene Symbol: LOC100911044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748732.1 | XP_003748780.1 | 6 | 1742 |
Set Membership: |
G-protein coupled receptor |