Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: CYP2A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000764.2 | NP_000755.2 | 1 | 704 | ||
NM_030589.2 | NP_085079.2 | 1 | 704 | |||
XM_017026383.1 | XP_016881872.1 | 1 | 3901 | |||
GenBank mRNA | BC117165.1 | - | 1 | 207 | 40125716 | |
BC143325.1 | - | 1 | 207 | 9051830 | ||
M33317.1 | - | 1 | 171 | |||
U22029.1 | - | 1 | 705 |
Gene Symbol: CYP2A7P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U22030.1 | - | 1 | 525 | ||
U22044.1 | - | 1 | 525 |