Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI1 SIRNA Assays ›
Gene Symbol:
ABI1
Gene Name:
abl interactor 1
Gene Aliases:
ABI-1, ABLBP4, E3B1, NAP1BP, SSH3BP, SSH3BP1
Chromosome Location:
Chr.10: 26746596 - 26861087 on Build GRCh38
UniGene ID:
Hs.508148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19446
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012750.2
NP_001012768.1
3
636
NM_001012751.2
NP_001012769.1
3
636
NM_001012752.2
NP_001012770.1
3
636
NM_001178116.1
NP_001171587.1
4
687
NM_001178119.1
NP_001171590.1
3
636
NM_001178120.1
NP_001171591.1
3
636
NM_001178121.1
NP_001171592.1
3
636
NM_001178122.1
NP_001171593.1
3
636
NM_001178123.1
NP_001171594.1
3
636
NM_001178124.1
NP_001171595.1
3
636
NM_005470.3
NP_005461.2
3
636
XM_005252332.3
XP_005252389.1
3
451
XM_005252333.3
XP_005252390.1
3
451
XM_006717359.2
XP_006717422.1
3
535
XM_006717360.2
XP_006717423.1
3
535
XM_006717361.2
XP_006717424.1
3
451
XM_011519289.2
XP_011517591.1
3
535
XM_017015458.1
XP_016870947.1
3
535
XM_017015459.1
XP_016870948.1
3
535
GenBank mRNA
AB040151.1
-
3
579
AB209268.1
-
3
487
AF001628.1
-
3
460
AF006516.1
-
3
478
AF260262.1
-
3
535
AF540955.1
-
3
424
AK126803.1
-
4
586
AK291823.1
-
3
497
AK296751.1
-
2
201
BC013238.1
-
3
484
BC024254.2
-
3
487
3531592
FJ380057.1
-
3
412
FJ380058.1
-
3
412
FJ380059.1
-
3
412
U87166.1
-
3
493
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
somitogenesis
movement of cell or subcellular component
transmembrane receptor protein tyrosine kinase signaling pathway
actin polymerization or depolymerization
negative regulation of cell proliferation
viral process
peptidyl-tyrosine phosphorylation
megakaryocyte development
Fc-gamma receptor signaling pathway involved in phagocytosis
vascular endothelial growth factor receptor signaling pathway
positive regulation of protein tyrosine kinase activity
lamellipodium morphogenesis
cell-cell adhesion
protein binding
cytoskeletal protein binding
protein tyrosine kinase activator activity
protein complex binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0