Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GUK1 SIRNA Assays ›
          Gene Symbol:
          GUK1
          Gene Name:
          guanylate kinase 1
          Gene Aliases:
          GMK
          Chromosome Location:
          Chr.1: 228139643 - 228148955 on Build GRCh38
          UniGene ID:
          Hs.376933
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s194482
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GUK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000858.5 NP_000849.1 6 587
          NM_001159390.1 NP_001152862.1 5 422
          NM_001159391.1 NP_001152863.1 5 336
          NM_001242839.1 NP_001229768.1 5 362
          NM_001242840.1 NP_001229769.1 5 422
          XM_005273103.3 XP_005273160.1 7 758
          XM_005273104.4 XP_005273161.1 7 756
          XM_005273105.3 XP_005273162.1 6 615
          XM_005273106.2 XP_005273163.1 5 803
          XM_011544174.2 XP_011542476.1 5 3186
          XM_017001091.1 XP_016856580.1 7 740
          XM_017001092.1 XP_016856581.1 6 1075
          XM_017001093.1 XP_016856582.1 6 535
          XM_017001094.1 XP_016856583.1 5 373
          XM_017001095.1 XP_016856584.1 6 532
          XM_017001096.1 XP_016856585.1 5 372
          GenBank mRNA AK303845.1 - 5 375
          AL832986.1 - 2 2098
          BC006249.2 - 6 525 3946293
          BC007369.2 - 4 239
          BC009914.2 - 6 499 2821563
          BC091480.1 - 5 342 30340102
          BE780558.1 - 5 309
          BI771824.1 - 5 362
          BM790067.1 - 5 443
          BU194785.1 - 388
          CR456836.1 - 4 284
          CR542006.1 - 4 284
          L76200.1 - 4 284
          U66895.1 - 4 290

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00176133_m1

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide metabolic process
          dGDP biosynthetic process
          nucleobase-containing small molecule interconversion
          GDP-mannose metabolic process
          glycoprotein transport
          ATP metabolic process
          GMP metabolic process
          dGMP metabolic process
          dATP metabolic process
          GDP biosynthetic process
          nucleotide phosphorylation
          guanylate kinase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0