Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDCD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267556.1 | NP_001254485.1 | 4 | 431 | ||
NM_001267558.1 | NP_001254487.1 | 5 | 346 | |||
NM_013232.3 | NP_037364.1 | 4 | 431 | |||
GenBank mRNA | AF035606.1 | - | 4 | 457 | ||
AK001917.1 | - | 4 | 382 | |||
AK223366.1 | - | 4 | 431 | |||
AK308872.1 | - | 3 | 329 | |||
AK315370.1 | - | 4 | 431 | |||
BC012384.1 | - | 4 | 399 | 3859672 | ||
BC106706.2 | - | 4 | 380 | 40003290 | ||
BC110291.1 | - | 4 | 395 | 5187550 | ||
BI553617.1 | - | 4 | 433 | |||
BT020072.1 | - | 4 | 331 | |||
DB495449.1 | - | 5 | 331 | |||
U58773.1 | - | 4 | 362 |