Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L11 SIRNA Assays ›
Gene Symbol:
BCL2L11
Gene Name:
BCL2 like 11
Gene Aliases:
BAM, BIM, BOD
Chromosome Location:
Chr.2: 111120914 - 111168445 on Build GRCh38
UniGene ID:
Hs.469658
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19474
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204108.1
NP_001191037.1
2
614
NM_001204109.1
NP_001191038.1
2
614
NM_001204111.1
NP_001191040.1
3
434
NM_001204112.1
NP_001191041.1
3
434
NM_001204113.1
NP_001191042.1
3
434
NM_006538.4
NP_006529.1
3
434
NM_138621.4
NP_619527.1
2
614
NM_138622.3
NP_619528.1
2
614
NM_138623.3
NP_619529.1
3
434
NM_138624.3
NP_619530.1
2
614
NM_138626.3
NP_619532.1
2
614
NM_207002.3
NP_996885.1
3
434
XM_005263550.3
XP_005263607.1
2
1392
XM_005263551.2
XP_005263608.1
2
1392
XM_005263552.3
XP_005263609.1
3
1212
XM_005263553.2
XP_005263610.1
3
1212
XM_005263554.2
XP_005263611.1
2
1392
XM_005263556.3
XP_005263613.1
2
614
XM_005263557.4
XP_005263614.1
2
691
XM_005263561.3
XP_005263618.1
3
434
XM_011510461.2
XP_011508763.1
2
1392
XM_011510464.2
XP_011508766.1
3
1212
XM_017003101.1
XP_016858590.1
3
1212
GenBank mRNA
AB071195.1
-
1
326
AB071196.1
-
2
146
AB071197.1
-
1
326
AB071198.1
-
1
326
AF032457.1
-
1
326
AF032458.1
-
2
146
AK290377.1
-
3
434
AK291269.1
-
2
611
AY352518.1
-
2
146
AY423443.1
-
1
326
AY428962.1
-
1
326
BC033694.1
-
2
596
5213713
CN411912.1
-
2
583
DQ849201.1
-
2
146
DQ849202.1
-
2
146
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
B cell homeostasis
B cell apoptotic process
kidney development
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
myeloid cell homeostasis
apoptotic process
meiosis I
cell-matrix adhesion
spermatogenesis
brain development
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
mammary gland development
positive regulation of protein homooligomerization
response to endoplasmic reticulum stress
tube formation
odontogenesis of dentin-containing tooth
T cell homeostasis
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ear development
positive regulation of cell cycle
regulation of organ growth
developmental pigmentation
regulation of developmental pigmentation
spleen development
thymus development
post-embryonic organ morphogenesis
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
apoptotic process involved in embryonic digit morphogenesis
positive regulation of IRE1-mediated unfolded protein response
positive regulation of fibroblast apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
protein binding
microtubule binding
dynein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0