Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2L11 SIRNA Assays ›
          Gene Symbol:
          BCL2L11
          Gene Name:
          BCL2 like 11
          Gene Aliases:
          BAM, BIM, BOD
          Chromosome Location:
          Chr.2: 111120914 - 111168445 on Build GRCh38
          UniGene ID:
          Hs.469658
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s195012

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL2L11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204106.1 NP_001191035.1 2 332
          NM_001204107.1 NP_001191036.1 2 332
          NM_001204108.1 NP_001191037.1 2 332
          NM_001204109.1 NP_001191038.1 2 332
          NM_001204110.1 NP_001191039.1 2 332
          NM_001204111.1 NP_001191040.1 2 332
          NM_001204112.1 NP_001191041.1 2 332
          NM_001204113.1 NP_001191042.1 2 332
          NM_006538.4 NP_006529.1 2 332
          NM_138621.4 NP_619527.1 2 332
          NM_138622.3 NP_619528.1 2 332
          NM_138623.3 NP_619529.1 2 332
          NM_138624.3 NP_619530.1 2 332
          NM_138625.3 NP_619531.1 2 332
          NM_138626.3 NP_619532.1 2 332
          NM_138627.3 NP_619533.1 2 332
          NM_207002.3 NP_996885.1 2 332
          NM_207003.2 NP_996886.1 2 332
          XM_005263550.3 XP_005263607.1 2 1110
          XM_005263551.2 XP_005263608.1 2 1110
          XM_005263552.3 XP_005263609.1 2 1110
          XM_005263553.2 XP_005263610.1 2 1110
          XM_005263554.2 XP_005263611.1 2 1110
          XM_005263555.3 XP_005263612.1 2 1110
          XM_005263556.3 XP_005263613.1 2 332
          XM_005263557.4 XP_005263614.1 2 409
          XM_005263559.2 XP_005263616.1 2 1110
          XM_005263561.3 XP_005263618.1 2 332
          XM_011510461.2 XP_011508763.1 2 1110
          XM_011510464.2 XP_011508766.1 2 1110
          XM_017003101.1 XP_016858590.1 2 1110
          XM_017003102.1 XP_016858591.1 2 332
          GenBank mRNA AB071195.1 - 1 44
          AB071196.1 - 1 44
          AB071197.1 - 1 44
          AB071198.1 - 1 44
          AB071199.1 - 1 44
          AB071200.1 - 1 44
          AF032457.1 - 1 44
          AF032458.1 - 1 44
          AK290377.1 - 2 332
          AK291269.1 - 2 329
          AY305714.1 - 1 44
          AY305715.1 - 1 44
          AY352518.1 - 1 44
          AY423441.1 - 1 44
          AY423442.1 - 1 44
          AY423443.1 - 1 44
          AY428962.1 - 1 44
          BC033694.1 - 2 314 5213713
          CN411912.1 - 2 301
          DQ849200.1 - 1 44
          DQ849201.1 - 1 44
          DQ849202.1 - 1 44

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          B cell homeostasis
          B cell apoptotic process
          kidney development
          protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          myeloid cell homeostasis
          apoptotic process
          meiosis I
          cell-matrix adhesion
          spermatogenesis
          brain development
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          mammary gland development
          positive regulation of protein homooligomerization
          response to endoplasmic reticulum stress
          tube formation
          odontogenesis of dentin-containing tooth
          T cell homeostasis
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of neuron apoptotic process
          ear development
          positive regulation of cell cycle
          regulation of organ growth
          developmental pigmentation
          regulation of developmental pigmentation
          spleen development
          thymus development
          post-embryonic organ morphogenesis
          positive regulation of apoptotic process by virus
          cellular process regulating host cell cycle in response to virus
          thymocyte apoptotic process
          positive regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          apoptotic process involved in embryonic digit morphogenesis
          positive regulation of IRE1-mediated unfolded protein response
          positive regulation of fibroblast apoptotic process
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          microtubule binding
          dynein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0