Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNAJB6 SIRNA Assays ›
          Gene Symbol:
          DNAJB6 TMEM135
          Gene Name:
          DnaJ heat shock protein family (Hsp40) member B6
          transmembrane protein 135
          Gene Aliases:
          DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
          PMP52
          Chromosome Location:
          Chr.7: 157337004 - 157417439 on Build GRCh38
          Chromosome Location:
          Chr.11: 87037844 - 87328834 on Build GRCh38
          UniGene ID:
          Hs.490745
          Hs.188591
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s19532

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DNAJB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005494.2 NP_005485.1 8 793
          NM_058246.3 NP_490647.1 8 792
          XM_005249515.2 XP_005249572.1 8 784
          XM_005249516.2 XP_005249573.1 8 928
          XM_006715823.1 XP_006715886.1 8 784
          XM_011515704.1 XP_011514006.1 8 763
          GenBank mRNA AB014888.1 - 8 738
          AB015798.1 - 8 785
          AB015799.1 - 8 785
          AB209859.1 - 8 786
          AF060703.1 - 8 783
          AF075601.1 - 8 770
          AK124827.1 - 3 1815
          AK223601.1 - 8 803
          AK291953.1 - 8 775
          AK293025.1 - 8 776
          AK297796.1 - 7 683
          AK297989.1 - 8 652
          AK301152.1 - 6 631
          AL136707.1 - 8 797
          AL832124.1 - 3 4409
          BC000177.2 - 8 784 2900421
          BC002446.2 - 8 792 3346070
          BC043270.1 - 3 2504
          CR533498.1 - 7 638
          EU075177.1 - 8 782
          Gene Symbol: TMEM135
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AF080569.1 - 8 692

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001037940, NM_001127367, NM_011847, AK153091, AK152626, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          actin cytoskeleton organization
          extracellular matrix organization
          positive regulation of ATPase activity
          regulation of protein localization
          protein localization to nucleus
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          intermediate filament organization
          negative regulation of transcription, DNA-templated
          chorio-allantoic fusion
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          chorion development
          negative regulation of inclusion body assembly
          regulation of cellular response to heat
          ATPase activator activity
          DNA binding
          protein binding
          heat shock protein binding
          unfolded protein binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0