Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF655 SIRNA Assays ›
Gene Symbol:
ZNF655
NUDCD3
Gene Name:
zinc finger protein 655
NudC domain containing 3
Gene Aliases:
VIK, VIK-1
NudCL
Chromosome Location:
Chr.7: 99557726 - 99576453 on Build GRCh38
Chromosome Location:
Chr.7: 44382366 - 44490875 on Build GRCh38
UniGene ID:
Hs.599798
Hs.488171
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s195374
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF655
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009958.1
NP_001009958.1
2
266
NM_001009960.1
NP_001009960.1
2
266
NM_001083956.1
NP_001077425.1
2
439
NM_001085366.1
NP_001078835.1
2
439
NM_001085367.1
NP_001078836.1
2
266
NM_001085368.1
NP_001078837.1
2
266
NM_024061.3
NP_076966.1
2
439
NM_138494.2
NP_612503.1
2
439
XM_017012598.1
XP_016868087.1
2
1126
XM_017012599.1
XP_016868088.1
2
284
XM_017012600.1
XP_016868089.1
2
297
XM_017012601.1
XP_016868090.1
2
283
XM_017012602.1
XP_016868091.1
2
1126
XM_017012603.1
XP_016868092.1
2
1359
XM_017012604.1
XP_016868093.1
2
276
XM_017012605.1
XP_016868094.1
2
160
XM_017012606.1
XP_016868095.1
2
297
XM_017012607.1
XP_016868096.1
2
283
XM_017012608.1
XP_016868097.1
2
1125
XM_017012609.1
XP_016868098.1
2
266
XM_017012610.1
XP_016868099.1
2
247
XM_017012611.1
XP_016868100.1
2
1135
XM_017012612.1
XP_016868101.1
2
1132
XM_017012613.1
XP_016868102.1
1
73
GenBank mRNA
AK027114.1
-
2
257
AK057245.1
-
3
165
AK225976.1
-
2
192
AK304785.1
-
2
143
AK314593.1
-
2
282
AK316024.1
-
2
257
AY099353.1
-
2
265
BC000823.1
-
2
209
BC004288.1
-
2
179
BC007378.2
-
2
133
BC037407.1
-
2
229
5752559
BC063478.1
-
2
439
BE293580.1
-
2
266
BI548533.1
-
2
186
BI912391.1
-
2
185
BM829333.1
-
2
293
BX490067.1
-
2
195
DN993232.1
-
2
202
Gene Symbol: NUDCD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK127163.1
-
139
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001008362, XM_008768981, XM_006248895, XM_003751126, XM_006248899, and others.
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0