Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
Gene Name:
BCL2 like 12
Gene Aliases:
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195442
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
7,898.00
Your Price
Online offer:
7898.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
7
1710
NM_001282516.1
NP_001269445.1
6
1440
NM_001282517.1
NP_001269446.1
5
1297
NM_001282519.1
NP_001269448.1
6
1489
NM_001282520.1
NP_001269449.1
5
1348
NM_001282521.1
NP_001269450.1
4
1261
NM_138639.1
NP_619580.1
7
1713
NR_104200.1
6
1570
NR_104201.1
7
1762
NR_104202.1
6
1619
NR_104203.1
5
1346
NR_104204.1
4
1205
NR_104205.1
3
1118
XM_017027345.1
XP_016882834.1
6
1507
GenBank mRNA
BC007724.2
-
7
848
BC104004.1
-
7
1286
40024338
BC104005.1
-
7
1289
40024342
BC104006.1
-
7
1289
40024345
KR820244.1
-
3
1502
KR820245.1
-
5
1867
KR820246.1
-
4
1594
KR820247.1
-
4
1643
KR820248.1
-
5
1916
KR820249.1
-
3
1551
KR820250.1
-
5
1916
KR820251.1
-
4
1643
KR820252.1
-
4
1692
KR820253.1
-
5
1965
KR820254.1
-
4
1283
KR820255.1
-
5
1375
KR820256.1
-
5
1424
KR820257.1
-
6
1697
KR820258.1
-
4
1332
KR820259.1
-
6
1697
KR820260.1
-
5
1424
KR820261.1
-
5
1473
KR820262.1
-
6
1746
KR820263.1
-
4
1280
KR820264.1
-
5
1372
KR820265.1
-
5
1421
KR820266.1
-
4
1329
KR820267.1
-
6
1694
KR820268.1
-
5
1421
KR820269.1
-
5
1470
KR820270.1
-
4
885
KR820271.1
-
6
1250
KR820272.1
-
5
977
KR820273.1
-
5
1026
KR820274.1
-
4
934
KR820275.1
-
6
1299
KR820276.1
-
5
1026
KR820277.1
-
5
1075
KR820278.1
-
5
715
KR820279.1
-
7
1080
KR820280.1
-
6
807
KR820281.1
-
6
856
KR820282.1
-
6
755
KR820283.1
-
6
804
KR820284.1
-
5
712
KR820285.1
-
7
1077
KR820286.1
-
6
804
KR820287.1
-
6
853
KR820288.1
-
3
1140
KR820289.1
-
4
1232
KR820290.1
-
4
1281
KR820291.1
-
5
1505
KR820292.1
-
5
1554
KR820293.1
-
2
1053
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00369327_m1
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0