Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
Gene Name:
BCL2 like 12
Gene Aliases:
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195443
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
1
846
NM_001282516.1
NP_001269445.1
1
846
NM_001282517.1
NP_001269446.1
1
846
NM_001282519.1
NP_001269448.1
1
846
NM_001282520.1
NP_001269449.1
1
846
NM_001282521.1
NP_001269450.1
1
846
NM_138639.1
NP_619580.1
1
846
NR_104200.1
1
846
NR_104201.1
1
846
NR_104202.1
1
846
NR_104203.1
1
846
NR_104204.1
1
846
NR_104205.1
1
846
XM_017027345.1
XP_016882834.1
1
916
XM_017027346.1
XP_016882835.1
1
916
GenBank mRNA
BC104004.1
-
1
422
40024338
BC104005.1
-
1
422
40024342
BC104006.1
-
1
422
40024345
FJ868798.1
-
1
164
FJ868799.1
-
1
164
FJ868800.1
-
1
164
FJ868801.1
-
1
164
FJ868802.1
-
1
164
FJ868803.1
-
1
164
FJ911906.1
-
1
164
FJ911907.1
-
1
164
FJ911908.1
-
1
164
HM347053.1
-
1
164
KR820244.1
-
1
164
KR820245.1
-
1
164
KR820246.1
-
1
164
KR820247.1
-
1
164
KR820248.1
-
1
164
KR820249.1
-
1
164
KR820250.1
-
1
164
KR820251.1
-
1
164
KR820252.1
-
1
164
KR820253.1
-
1
164
KR820254.1
-
1
164
KR820255.1
-
1
164
KR820256.1
-
1
164
KR820257.1
-
1
164
KR820258.1
-
1
164
KR820259.1
-
1
164
KR820260.1
-
1
164
KR820261.1
-
1
164
KR820262.1
-
1
164
KR820263.1
-
1
164
KR820264.1
-
1
164
KR820265.1
-
1
164
KR820266.1
-
1
164
KR820267.1
-
1
164
KR820268.1
-
1
164
KR820269.1
-
1
164
KR820270.1
-
1
164
KR820271.1
-
1
164
KR820272.1
-
1
164
KR820273.1
-
1
164
KR820274.1
-
1
164
KR820275.1
-
1
164
KR820276.1
-
1
164
KR820277.1
-
1
164
KR820278.1
-
1
164
KR820279.1
-
1
164
KR820280.1
-
1
164
KR820281.1
-
1
164
KR820282.1
-
1
164
KR820283.1
-
1
164
KR820284.1
-
1
164
KR820285.1
-
1
164
KR820286.1
-
1
164
KR820287.1
-
1
164
KR820288.1
-
1
164
KR820289.1
-
1
164
KR820290.1
-
1
164
KR820291.1
-
1
164
KR820292.1
-
1
164
KR820293.1
-
1
164
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fpbqh:80/100.66.79.145:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0