Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT3A SIRNA Assays ›
Gene Symbol:
WNT3A
Gene Name:
Wnt family member 3A
Gene Aliases:
-
Chromosome Location:
Chr.1: 228007022 - 228067113 on Build GRCh38
UniGene ID:
Hs.336930
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195525
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNT3A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033131.3
NP_149122.1
4
1876
GenBank mRNA
AK056278.1
-
4
1847
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
in utero embryonic development
positive regulation of cytokine production
positive regulation of protein phosphorylation
heart looping
positive regulation of receptor internalization
negative regulation of heart induction by canonical Wnt signaling pathway
axon guidance
synapse assembly
positive regulation of cell proliferation
COP9 signalosome assembly
negative regulation of neuron projection development
Wnt signaling pathway
spinal cord association neuron differentiation
hippocampus development
cell proliferation in forebrain
Wnt signaling pathway involved in forebrain neuroblast division
dorsal/ventral neural tube patterning
hemopoiesis
neuron differentiation
extracellular matrix organization
mammary gland development
positive regulation of B cell proliferation
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
cell proliferation in midbrain
cellular protein localization
positive regulation of catenin import into nucleus
skeletal muscle cell differentiation
post-anal tail morphogenesis
synaptic vesicle recycling
inner ear morphogenesis
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of mesodermal cell fate specification
paraxial mesodermal cell fate commitment
positive regulation of skeletal muscle tissue development
positive regulation of collateral sprouting in absence of injury
negative regulation of axon extension involved in axon guidance
negative regulation of neurogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
palate development
canonical Wnt signaling pathway
positive regulation of protein tyrosine kinase activity
positive regulation of dermatome development
canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
regulation of microtubule cytoskeleton organization
platelet aggregation
cellular response to retinoic acid
axis elongation involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
positive regulation of protein localization to plasma membrane
negative regulation of dopaminergic neuron differentiation
beta-catenin destruction complex disassembly
Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
positive regulation of cell-cell adhesion mediated by cadherin
positive regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
positive regulation of neural precursor cell proliferation
positive regulation of hepatocyte proliferation
positive regulation of transcription regulatory region DNA binding
positive regulation of cardiac muscle cell differentiation
transcription coactivator activity
receptor binding
frizzled binding
frizzled-2 binding
protein binding
protein domain specific binding
co-receptor binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0