Search
Search

GNG8| Gene Symbol: GNG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033258.1 | NP_150283.1 | 1, 2 | 77 | ||
| XM_011527524.2 | XP_011525826.1 | 2, 3 | 637 | |||
| XM_017027503.1 | XP_016882992.1 | 2, 3 | 216 | |||
| XM_017027504.1 | XP_016882993.1 | 2, 3 | 2871 | |||
| XM_017027505.1 | XP_016882994.1 | 2, 3 | 170 | |||
| GenBank mRNA | AF493875.1 | - | 1, 2 | 77 | ||
| BC095514.1 | - | 1, 2 | 80 | 6519176 | ||