Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNM1L SIRNA Assays ›
          Gene Symbol:
          DNM1L
          Gene Name:
          dynamin 1 like
          Gene Aliases:
          DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV
          Chromosome Location:
          Chr.12: 32679200 - 32745650 on Build GRCh38
          UniGene ID:
          Hs.556296
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19559
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DNM1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278463.1 NP_001265392.1 6 687
          NM_001278464.1 NP_001265393.1 7 726
          NM_001278465.1 NP_001265394.1 7 726
          NM_005690.4 NP_005681.2 6 687
          NM_012062.4 NP_036192.2 6 687
          NM_012063.3 NP_036193.2 6 687
          XM_005253282.4 XP_005253339.1 7 698
          XM_011520543.2 XP_011518845.1 7 698
          GenBank mRNA AB006965.1 - 6 558
          AB209070.1 - 7 603
          AF000430.1 - 6 595
          AF061795.1 - 6 544
          AF151685.1 - 6 544
          AK291094.1 - 6 567
          AK293290.1 - 5 480
          AK299926.1 - 7 639
          AK302565.1 - 6 687
          AK311606.1 - 6 682
          BC024590.1 - 6 557 3882922
          BC107763.1 - 6 571

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00247147_m1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          release of cytochrome c from mitochondria
          dynamin polymerization involved in mitochondrial fission
          endocytosis
          cellular component disassembly involved in execution phase of apoptosis
          regulation of mitochondrion organization
          peroxisome fission
          regulation of protein oligomerization
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          mitochondrial fragmentation involved in apoptotic process
          positive regulation of protein secretion
          protein oligomerization
          protein homotetramerization
          heart contraction
          membrane fusion
          necroptotic process
          mitochondrion morphogenesis
          protein localization to mitochondrion
          positive regulation of mitochondrial fission
          mitochondrial membrane fission
          positive regulation of release of cytochrome c from mitochondria
          regulation of peroxisome organization
          regulation of mitophagy
          regulation of ATP metabolic process
          positive regulation of intrinsic apoptotic signaling pathway
          GTPase activity
          GTPase activator activity
          protein binding
          GTP binding
          lipid binding
          Rab GTPase binding
          GTP-dependent protein binding
          ubiquitin protein ligase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0