Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 9 | 1185 | ||
XM_006721179.1 | XP_006721242.1 | 9 | 1281 | |||
XM_011523034.1 | XP_011521336.1 | 10 | 1527 | |||
XM_011523036.2 | XP_011521338.1 | 8 | 1111 | |||
XM_011523037.1 | XP_011521339.1 | 8 | 1174 | |||
GenBank mRNA | AF448856.1 | - | 9 | 1335 | ||
AL833339.1 | - | 7 | 2283 | |||
BC039068.1 | - | 8 | 1967 | 5188643 |
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 10 | 1217 | ||
NM_001271197.1 | NP_001258126.1 | 10 | 1297 | |||
NM_182619.3 | NP_872425.2 | 10 | 1323 | |||
XM_005255934.1 | XP_005255991.1 | 9 | 1266 | |||
XM_017023206.1 | XP_016878695.1 | 9 | 1266 | |||
XM_017023207.1 | XP_016878696.1 | 8 | 1096 | |||
XM_017023208.1 | XP_016878697.1 | 9 | 1449 | |||
XM_017023209.1 | XP_016878698.1 | 8 | 1159 | |||
GenBank mRNA | AF428259.1 | - | 9 | 1335 | ||
AF521893.1 | - | 10 | 1336 | |||
AK074773.1 | - | 9 | 1151 | |||
AK289884.1 | - | 9 | 1265 | |||
BC078143.1 | - | 10 | 1199 | 5761001 | ||
BC141808.1 | - | 10 | 1297 | 40147524 |
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 9 | 1279 | ||
XM_005255976.2 | XP_005256033.1 | 9 | 1356 | |||
XM_011523093.2 | XP_011521395.1 | 8 | 1062 | |||
XM_017023255.1 | XP_016878744.1 | 10 | 1533 | |||
XM_017023256.1 | XP_016878745.1 | 8 | 1249 | |||
GenBank mRNA | BM695392.1 | - | 5 | 490 |