Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBMY1A1 SIRNA Assays ›
          Gene Symbol:
          RBMY1A1 RBMY1B RBMY1D RBMY1E RBMY1F RBMY1J
          Gene Name:
          RNA binding motif protein, Y-linked, family 1, member A1
          RNA binding motif protein, Y-linked, family 1, member B
          RNA binding motif protein, Y-linked, family 1, member D
          RNA binding motif protein, Y-linked, family 1, member E
          RNA binding motif protein, Y-linked, family 1, member F
          RNA binding motif protein, Y-linked, family 1, member J
          Gene Aliases:
          RBM, RBM1, RBM2, RBMY, RBMY1C, YRRM1, YRRM2
          -
          -
          -
          YRRM2
          -
          Chromosome Location:
          Chr.Y: 21534879 - 21559683 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21511338 - 21527212 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21869719 - 21900746 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21902174 - 21918044 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22167099 - 22182942 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22403410 - 22419317 on Build GRCh38
          UniGene ID:
          Hs.380450
          Hs.380450
          Hs.380450
          Hs.725744
          Hs.567746
          Hs.380450
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s196162

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBMY1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320944.1 NP_001307873.1 4 451
          NM_005058.3 NP_005049.1 4 451
          XM_011531437.2 XP_011529739.1 4 390
          XM_011531438.1 XP_011529740.1 4 390
          XM_017030060.1 XP_016885549.1 3 227
          GenBank mRNA BC070298.1 - 4 416 6615329
          BC146853.1 - 4 364 9056781
          BU568309.1 - 4 419
          X76059.1 - 4 416
          Gene Symbol: RBMY1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006121.3 NP_001006121.1 4 451
          NM_001320948.1 NP_001307877.1 4 451
          XM_011531433.1 XP_011529735.1 4 411
          XM_017030056.1 XP_016885545.1 4 411
          XM_017030057.1 XP_016885546.1 3 227
          GenBank mRNA BC136862.1 - 4 364 9020853
          BC140758.1 - 4 364 9021626
          BC144459.1 - 4 364 9052985
          Gene Symbol: RBMY1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006120.3 NP_001006120.2 4 451
          NM_001320949.1 NP_001307878.1 4 451
          NM_001320950.1 NP_001307879.1 5 502
          XM_011531486.2 XP_011529788.1 4 324
          XM_011531487.2 XP_011529789.1 4 411
          XM_011531488.2 XP_011529790.1 4 411
          XM_011531489.1 XP_011529791.1 4 411
          GenBank mRNA BC144422.1 - 4 364 9052943
          BC144423.1 - 5 415
          Gene Symbol: RBMY1E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006118.2 NP_001006118.2 4 411
          XM_011531493.1 XP_011529795.1 4 411
          XM_011531494.2 XP_011529796.1 4 411
          XM_011531495.1 XP_011529797.1 4 411
          XM_017030058.1 XP_016885547.1 3 227
          Gene Symbol: RBMY1F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303410.1 NP_001290339.1 4 411
          NM_152585.2 NP_689798.1 4 411
          XM_017030033.1 XP_016885522.1 3 227
          GenBank mRNA BC030018.2 - 4 411 5269012
          X76060.1 - 4 393
          Gene Symbol: RBMY1J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006117.3 NP_001006117.2 4 476
          XM_011531500.1 XP_011529802.1 4 347
          XM_011531502.1 XP_011529804.1 4 347
          XM_017030059.1 XP_016885548.1 3 227
          GenBank mRNA AK313597.1 - 4 480
          BC144577.1 - 4 364 9053106
          BC152392.1 - 3 321 8860211

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          spermatogenesis
          RNA splicing
          male gonad development
          nucleotide binding
          RNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7q72q:80/100.66.76.177:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0