Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBMY1B SIRNA Assays ›
          Gene Symbol:
          RBMY1B RBMY1A1 RBMY1A3P RBMY1D RBMY1E RBMY1F RBMY1J RBMY2FP
          Gene Name:
          RNA binding motif protein, Y-linked, family 1, member B
          RNA binding motif protein, Y-linked, family 1, member A1
          RNA binding motif protein, Y-linked, family 1, member A3 pseudogene
          RNA binding motif protein, Y-linked, family 1, member D
          RNA binding motif protein, Y-linked, family 1, member E
          RNA binding motif protein, Y-linked, family 1, member F
          RNA binding motif protein, Y-linked, family 1, member J
          RNA binding motif protein, Y-linked, family 2, member F pseudogene
          Gene Aliases:
          -
          RBM, RBM1, RBM2, RBMY, RBMY1C, YRRM1, YRRM2
          -
          -
          -
          YRRM2
          -
          -
          Chromosome Location:
          Chr.Y: 21511338 - 21527212 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21534879 - 21559683 on Build GRCh38
          Chromosome Location:
          Chr.Y: 9317061 - 9322874 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21869719 - 21900746 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21902174 - 21918044 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22167099 - 22182942 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22403410 - 22419317 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22308859 - 22316205 on Build GRCh38
          UniGene ID:
          Hs.380450
          Hs.380450
          Hs.567833
          Hs.380450
          Hs.725744
          Hs.567746
          Hs.380450
          Hs.711527
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s196166

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBMY1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006121.3 NP_001006121.1 4 416
          NM_001320948.1 NP_001307877.1 4 416
          XM_011531433.1 XP_011529735.1 4 376
          XM_017030056.1 XP_016885545.1 4 376
          XM_017030057.1 XP_016885546.1 3 192
          GenBank mRNA BC136862.1 - 4 329 9020853
          BC140758.1 - 4 329 9021626
          BC144459.1 - 4 329 9052985
          Gene Symbol: RBMY1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320944.1 NP_001307873.1 4 416
          NM_005058.3 NP_005049.1 4 416
          XM_011531437.2 XP_011529739.1 4 355
          XM_011531438.1 XP_011529740.1 4 355
          XM_017030060.1 XP_016885549.1 3 192
          GenBank mRNA BC070298.1 - 4 381 6615329
          BC146853.1 - 4 329 9056781
          BU568309.1 - 4 384
          X76059.1 - 4 381
          Gene Symbol: RBMY1A3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_001547.1 3 269
          Gene Symbol: RBMY1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006120.3 NP_001006120.2 4 416
          NM_001320949.1 NP_001307878.1 4 416
          NM_001320950.1 NP_001307879.1 5 467
          XM_011531486.2 XP_011529788.1 4 289
          XM_011531487.2 XP_011529789.1 4 376
          XM_011531488.2 XP_011529790.1 4 376
          XM_011531489.1 XP_011529791.1 4 376
          GenBank mRNA BC144422.1 - 4 329 9052943
          BC144423.1 - 5 380
          Gene Symbol: RBMY1E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006118.2 NP_001006118.2 4 376
          XM_011531493.1 XP_011529795.1 4 376
          XM_011531494.2 XP_011529796.1 4 376
          XM_011531495.1 XP_011529797.1 4 376
          XM_017030058.1 XP_016885547.1 3 192
          Gene Symbol: RBMY1F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303410.1 NP_001290339.1 4 376
          NM_152585.2 NP_689798.1 4 376
          XM_017030033.1 XP_016885522.1 3 192
          GenBank mRNA BC030018.2 - 4 376 5269012
          X76060.1 - 4 358
          Gene Symbol: RBMY1J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006117.3 NP_001006117.2 4 441
          XM_011531500.1 XP_011529802.1 4 312
          XM_011531502.1 XP_011529804.1 4 312
          XM_017030059.1 XP_016885548.1 3 192
          GenBank mRNA AK313597.1 - 4 445
          BC144577.1 - 4 329 9053106
          BC152392.1 - 3 286 8860211
          Gene Symbol: RBMY2FP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002193.2 4 380
          GenBank mRNA BC026077.2 - 4 382

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          spermatogenesis
          RNA splicing
          male gonad development
          nucleotide binding
          RNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0