Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005715.2 | NP_005706.1 | 7 | 1110 | ||
GenBank mRNA | AB020316.1 | - | 7 | 917 | ||
AK292922.1 | - | 7 | 858 | |||
AK315320.1 | - | 7 | 818 | |||
BC093668.1 | - | 7 | 862 | 7939513 | ||
BC093694.1 | - | 7 | 862 | 7939539 |