Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABI2 SIRNA Assays ›
          Gene Symbol:
          ABI2
          Gene Name:
          abl-interactor 2
          Gene Aliases:
          ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
          Chromosome Location:
          Chr.2: 203327874 - 203432173 on Build GRCh38
          UniGene ID:
          Hs.471156
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19771

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABI2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282925.1 NP_001269854.1 6 908
          NM_001282926.1 NP_001269855.1 5 890
          NM_001282927.1 NP_001269856.1 5 566
          NM_001282932.1 NP_001269861.1 6 1087
          NM_005759.5 NP_005750.4 5 890
          XM_005246217.2 XP_005246274.1 6 1273
          XM_005246218.2 XP_005246275.1 5 1255
          XM_005246226.2 XP_005246283.1 5 1255
          XM_006712174.2 XP_006712237.1 6 1273
          XM_006712177.2 XP_006712240.1 7 1349
          XM_006712180.2 XP_006712243.1 6 1273
          XM_006712181.2 XP_006712244.1 6 1273
          XM_006712182.2 XP_006712245.1 6 1273
          XM_006712185.1 XP_006712248.1 6 1331
          XM_011510473.2 XP_011508775.1 5 572
          XM_017003118.1 XP_016858607.1 6 1273
          XM_017003119.1 XP_016858608.1 6 1273
          XM_017003120.1 XP_016858609.1 6 1273
          XM_017003121.1 XP_016858610.1 6 1273
          XM_017003124.1 XP_016858613.1 5 1255
          XM_017003125.1 XP_016858614.1 5 1255
          XM_017003126.1 XP_016858615.1 5 1255
          XM_017003127.1 XP_016858616.1 5 1255
          XM_017003128.1 XP_016858617.1 7 1349
          XM_017003132.1 XP_016858621.1 6 1273
          XM_017003134.1 XP_016858623.1 5 1255
          XM_017003135.1 XP_016858624.1 5 1255
          XM_017003136.1 XP_016858625.1 5 1255
          XM_017003137.1 XP_016858626.1 7 1349
          XM_017003139.1 XP_016858628.1 6 1331
          XM_017003140.1 XP_016858629.1 6 1331
          GenBank mRNA AF260261.1 - 6 632
          AK299672.1 - 4 198
          AK299824.1 - 5 734
          AK302853.1 - 6 1013
          BC001439.2 - 5 639 3138341
          BT009920.1 - 5 614
          U23435.1 - 5 527
          U31089.1 - 4 359
          X95632.1 - 5 714

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_173143, BC097273, FQ222350, U94904.
          This siRNA also targets the following transcripts in Mouse: NM_001198571, NM_001198570, NM_198127, BC079545, BC079646, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          cytoskeleton organization
          actin polymerization or depolymerization
          viral process
          cell migration
          Rac protein signal transduction
          peptidyl-tyrosine phosphorylation
          positive regulation of Arp2/3 complex-mediated actin nucleation
          DNA binding
          protein binding
          cytoskeletal adaptor activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          Rac GTPase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-h6x29:80/100.66.79.70:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0