Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: CCDC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258304.1 | NP_001245233.1 | 1 | 457 | ||
XM_005263835.3 | XP_005263892.1 | 1 | 429 | |||
XM_006712828.3 | XP_006712891.1 | 1 | 430 | |||
XM_017005234.1 | XP_016860723.1 | 1 | 431 | |||
XM_017005236.1 | XP_016860725.1 | 1 | 430 | |||
XM_017005237.1 | XP_016860726.1 | 1 | 446 | |||
XM_017005238.1 | XP_016860727.1 | 1 | 455 | |||
XM_017005242.1 | XP_016860731.1 | 1 | 438 | |||
XM_017005243.1 | XP_016860732.1 | 1 | 449 | |||
GenBank mRNA | AB299086.1 | - | 1 | 372 |
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263840.2 | XP_005263897.1 | 1 | 357 | ||
XM_005263842.2 | XP_005263899.1 | 1 | 357 | |||
XM_006712833.2 | XP_006712896.1 | 1 | 357 | |||
XM_006712835.2 | XP_006712898.1 | 1 | 357 | |||
XM_011512147.1 | XP_011510449.1 | 1 | 357 | |||
GenBank mRNA | AK297246.1 | - | 1 | 357 |