Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other USP17L6P SIRNA Assays ›
          Gene Symbol:
          USP17L6P LOC392196 USP17L1 USP17L10 USP17L11 USP17L12 USP17L13 USP17L15 USP17L17 USP17L18 USP17L19 USP17L2 USP17L20 USP17L21 USP17L22 USP17L24 USP17L25 USP17L26 USP17L27 USP17L28 USP17L29 USP17L3 USP17L30 USP17L4 USP17L5 USP17L8 USP17L9P
          Gene Name:
          ubiquitin specific peptidase 17-like family member 6, pseudogene
          ubiquitin specific peptidase 17-like family member 2 pseudogene
          ubiquitin specific peptidase 17-like family member 1
          ubiquitin specific peptidase 17-like family member 10
          ubiquitin specific peptidase 17-like family member 11
          ubiquitin specific peptidase 17-like family member 12
          ubiquitin specific peptidase 17-like family member 13
          ubiquitin specific peptidase 17-like family member 15
          ubiquitin specific peptidase 17-like family member 17
          ubiquitin specific peptidase 17-like family member 18
          ubiquitin specific peptidase 17-like family member 19
          ubiquitin specific peptidase 17-like family member 2
          ubiquitin specific peptidase 17-like family member 20
          ubiquitin specific peptidase 17-like family member 21
          ubiquitin specific peptidase 17-like family member 22
          ubiquitin specific peptidase 17-like family member 24
          ubiquitin specific peptidase 17-like family member 25
          ubiquitin specific peptidase 17-like family member 26
          ubiquitin specific peptidase 17-like family member 27
          ubiquitin specific peptidase 17-like family member 28
          ubiquitin specific peptidase 17-like family member 29
          ubiquitin specific peptidase 17-like family member 3
          ubiquitin specific peptidase 17-like family member 30
          ubiquitin specific peptidase 17-like family member 4
          ubiquitin specific peptidase 17-like family member 5
          ubiquitin specific peptidase 17-like family member 8
          ubiquitin specific peptidase 17-like family member 9, pseudogene
          Gene Aliases:
          DUB4, USP17L6, vDUB4
          -
          USP17L1P
          -
          -
          -
          -
          -
          -
          -
          -
          DUB-3, DUB3, USP17
          -
          -
          -
          -
          -
          -
          -
          -
          -
          USP17B, USP17F
          -
          -
          -
          USP17L10
          RS447, USP17, USP17A, USP17H, USP17I, USP17J, USP17K, USP17L, USP17M
          Chromosome Location:
          Chr.4: 9367874 - 9369070 on Build GRCh38
          Chromosome Location:
          Chr.8: 12127858 - 12129297 on Build GRCh38
          Chromosome Location:
          Chr.8: 7332387 - 7333979 on Build GRCh38
          Chromosome Location:
          Chr.4: 9210657 - 9212249 on Build GRCh38
          Chromosome Location:
          Chr.4: 9215405 - 9216997 on Build GRCh38
          Chromosome Location:
          Chr.4: 9220152 - 9221744 on Build GRCh38
          Chromosome Location:
          Chr.4: 9224896 - 9226488 on Build GRCh38
          Chromosome Location:
          Chr.4: 9234385 - 9236334 on Build GRCh38
          Chromosome Location:
          Chr.4: 9243879 - 9245471 on Build GRCh38
          Chromosome Location:
          Chr.4: 9248630 - 9250222 on Build GRCh38
          Chromosome Location:
          Chr.4: 9253378 - 9254970 on Build GRCh38
          Chromosome Location:
          Chr.8: 12137168 - 12138760 on Build GRCh38
          Chromosome Location:
          Chr.4: 9258124 - 9259716 on Build GRCh38
          Chromosome Location:
          Chr.4: 9262872 - 9264464 on Build GRCh38
          Chromosome Location:
          Chr.4: 9267619 - 9269211 on Build GRCh38
          Chromosome Location:
          Chr.4: 9325165 - 9326757 on Build GRCh38
          Chromosome Location:
          Chr.4: 9329911 - 9331503 on Build GRCh38
          Chromosome Location:
          Chr.4: 9334658 - 9336250 on Build GRCh38
          Chromosome Location:
          Chr.4: 9344148 - 9345740 on Build GRCh38
          Chromosome Location:
          Chr.4: 9348893 - 9350485 on Build GRCh38
          Chromosome Location:
          Chr.4: 9353638 - 9355230 on Build GRCh38
          Chromosome Location:
          Chr.8: 7976393 - 7977985 on Build GRCh38
          Chromosome Location:
          Chr.4: 9363129 - 9364721 on Build GRCh38
          Chromosome Location:
          Chr.8: 7337115 - 7338707 on Build GRCh38
          Chromosome Location:
          Chr.4: 9339403 - 9340995 on Build GRCh38
          Chromosome Location:
          Chr.8: 7971661 - 7973253 on Build GRCh38
          Chromosome Location:
          Chr.4: 9358382 - 9359974 on Build GRCh38
          UniGene ID:
          Hs.722658
          Hs.742129
          Hs.653091
          Hs.741130
          Hs.722783
          Hs.741128
          Hs.741127
          Hs.528621
          Hs.741129
          Hs.741138
          Hs.741137
          Hs.531448
          Hs.722783
          Hs.741136
          Hs.723053
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.741131
          Hs.553810
          Hs.741134
          Hs.740243
          Hs.741135
          Hs.553810
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s198533

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: USP17L6P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027279.1 1 256
          GenBank mRNA AY533200.1 - 1 256
          Gene Symbol: LOC392196
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003275.2 1 256
          Gene Symbol: USP17L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256873.1 NP_001243802.1 1 256
          Gene Symbol: USP17L10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256852.1 NP_001243781.1 1 256
          Gene Symbol: USP17L11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256854.1 NP_001243783.1 1 256
          Gene Symbol: USP17L12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256853.1 NP_001243782.1 1 256
          Gene Symbol: USP17L13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256855.1 NP_001243784.1 1 256
          Gene Symbol: USP17L15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256894.1 NP_001243823.1 1 256
          Gene Symbol: USP17L17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256857.1 NP_001243786.1 1 256
          Gene Symbol: USP17L18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256859.1 NP_001243788.1 1 256
          Gene Symbol: USP17L19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256860.1 NP_001243789.1 1 256
          Gene Symbol: USP17L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201402.2 NP_958804.2 1 256
          GenBank mRNA AY509884.1 - 1 256
          Gene Symbol: USP17L20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256861.1 NP_001243790.1 1 256
          GenBank mRNA AY188990.1 - 1 256
          Gene Symbol: USP17L21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256862.1 NP_001243791.1 1 256
          Gene Symbol: USP17L22
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256863.1 NP_001243792.1 1 256
          Gene Symbol: USP17L24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242327.1 NP_001229256.1 1 256
          Gene Symbol: USP17L25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242326.1 NP_001229255.1 1 256
          Gene Symbol: USP17L26
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242328.1 NP_001229257.1 1 256
          Gene Symbol: USP17L27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242330.1 NP_001229259.1 1 256
          Gene Symbol: USP17L28
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242331.1 NP_001229260.1 1 256
          Gene Symbol: USP17L29
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242332.1 NP_001229261.1 1 256
          Gene Symbol: USP17L3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256871.1 NP_001243800.1 1 256
          Gene Symbol: USP17L30
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256867.1 NP_001243796.1 1 256
          Gene Symbol: USP17L4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256874.1 NP_001243803.1 1 256
          Gene Symbol: USP17L5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242329.1 NP_001229258.1 1 256
          Gene Symbol: USP17L8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256872.1 NP_001243801.1 1 256
          Gene Symbol: USP17L9P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046416.1 1 257
          GenBank mRNA AF544011.1 - 1 256
          BC100991.2 - 1 256
          BC100992.2 - 1 256
          BC100993.1 - 1 259

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          protein deubiquitination
          apoptotic process
          cell cycle checkpoint
          mitotic cell cycle checkpoint
          negative regulation of protein processing
          regulation of cell migration
          negative regulation of histone deacetylation
          negative regulation of GTPase activity
          regulation of cell proliferation
          regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of defense response to virus by host
          protein K63-linked deubiquitination
          protein K48-linked deubiquitination
          CAAX-box protein processing
          negative regulation of protein targeting to membrane
          regulation of ruffle assembly
          positive regulation of MDA-5 signaling pathway
          positive regulation of RIG-I signaling pathway
          thiol-dependent ubiquitinyl hydrolase activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0