Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPANXA1 SIRNA Assays ›
Gene Symbol:
SPANXA1
SPANXA2
SPANXC
SPANXD
Gene Name:
sperm protein associated with the nucleus, X-linked, family member A1
SPANX family member A2
SPANX family member C
SPANX family member D
Gene Aliases:
CT11.1, NAP-X, SPAN-X, SPAN-Xa, SPAN-Xb, SPANX, SPANX-A
CT11.1, SPANX, SPANX-A, SPANXA
C, CT11.3, CTp11, SPANX-C
CT11.4, SPANX-C, SPANX-D, SPANX-E, SPANXE, dJ171K16.1
Chromosome Location:
Chr.X: 141583674 - 141584738 on Build GRCh38
Chromosome Location:
Chr.X: 141589708 - 141590772 on Build GRCh38
Chromosome Location:
Chr.X: 141241463 - 141242550 on Build GRCh38
Chromosome Location:
Chr.X: 141697401 - 141698739 on Build GRCh38
UniGene ID:
Hs.334464
Hs.334464
Hs.375036
Hs.375036
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID
s198747
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPANXA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013453.3
NP_038481.2
2
370
GenBank mRNA
AF098306.1
-
2
346
AJ238277.1
-
2
349
AK225988.1
-
2
369
BC054023.1
-
2
371
6648369
BC069393.1
-
2
346
7262682
BC069816.1
-
2
346
7262670
CB115288.1
-
2
370
Gene Symbol: SPANXA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_145662.3
NP_663695.1
2
370
GenBank mRNA
CB110020.1
-
2
370
CB113891.1
-
2
370
Gene Symbol: SPANXC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022661.3
NP_073152.2
2
403
GenBank mRNA
AA412605.1
-
2
366
Gene Symbol: SPANXD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_032417.3
NP_115793.1
2
644
GenBank mRNA
AJ457795.1
-
2
346
BC005382.1
-
2
403
3935644
BC062672.1
-
2
584
4047937
BC103961.1
-
2
343
40013988
BC121070.2
-
2
353
40119569
BC134418.1
-
2
353
40119568
BM554791.1
-
2
644
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
biological_process
protein binding
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System