Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SPANXA1 SIRNA Assays ›
          Gene Symbol:
          SPANXA1 SPANXA2 SPANXC SPANXD
          Gene Name:
          sperm protein associated with the nucleus, X-linked, family member A1
          SPANX family member A2
          SPANX family member C
          SPANX family member D
          Gene Aliases:
          CT11.1, NAP-X, SPAN-X, SPAN-Xa, SPAN-Xb, SPANX, SPANX-A
          CT11.1, SPANX, SPANX-A, SPANXA
          C, CT11.3, CTp11, SPANX-C
          CT11.4, SPANX-C, SPANX-D, SPANX-E, SPANXE, dJ171K16.1
          Chromosome Location:
          Chr.X: 141583674 - 141584738 on Build GRCh38
          Chromosome Location:
          Chr.X: 141589708 - 141590772 on Build GRCh38
          Chromosome Location:
          Chr.X: 141241463 - 141242550 on Build GRCh38
          Chromosome Location:
          Chr.X: 141697401 - 141698739 on Build GRCh38
          UniGene ID:
          Hs.334464
          Hs.334464
          Hs.375036
          Hs.375036
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s198748

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SPANXA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013453.3 NP_038481.2 2 331
          GenBank mRNA AF098306.1 - 2 307
          AJ238277.1 - 2 310
          AK225988.1 - 2 330
          AY920732.1 - 2 271
          AY920733.1 - 2 271
          AY920735.1 - 2 271
          AY920736.1 - 2 271
          AY920737.1 - 2 271
          AY920738.1 - 2 271
          AY920739.1 - 2 271
          AY920741.1 - 2 271
          AY920742.1 - 2 271
          AY920743.1 - 2 271
          AY920744.1 - 2 271
          AY920745.1 - 2 271
          AY920746.1 - 2 271
          BC054023.1 - 2 332 6648369
          BC069393.1 - 2 307 7262682
          BC069816.1 - 2 307 7262670
          CB115288.1 - 2 331
          Gene Symbol: SPANXA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_145662.3 NP_663695.1 2 331
          GenBank mRNA CB110020.1 - 2 331
          CB113891.1 - 2 331
          Gene Symbol: SPANXC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022661.3 NP_073152.2 2 364
          GenBank mRNA AA412605.1 - 2 327
          Gene Symbol: SPANXD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032417.3 NP_115793.1 2 605
          GenBank mRNA AF312765.1 - 2 281
          AJ457795.1 - 2 307
          AY920734.1 - 2 271
          AY920740.1 - 2 271
          BC005382.1 - 2 364 3935644
          BC062672.1 - 2 545 4047937
          BC103961.1 - 2 304 40013988
          BC121070.2 - 2 314 40119569
          BC134418.1 - 2 314 40119568
          BM554791.1 - 2 605

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          biological_process
          protein binding
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-t2sjj:80/100.66.73.10:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0