Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCL3L3 SIRNA Assays ›
Gene Symbol:
CCL3L3
CCL3
CCL3L1
Gene Name:
C-C motif chemokine ligand 3 like 3
C-C motif chemokine ligand 3
C-C motif chemokine ligand 3 like 1
Gene Aliases:
464.2, D17S1718, G0S19-2, LD78, LD78BETA, SCYA3L, SCYA3L1
G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
464.2, D17S1718, G0S19-2, LD78, LD78-beta(1-70), LD78BETA, MIP1AP, SCYA3L, SCYA3L1
Chromosome Location:
Chr.17: 36194869 - 36196748 on Build GRCh38
Chromosome Location:
Chr.17: 36088256 - 36090160 on Build GRCh38
Chromosome Location:
-
UniGene ID:
Hs.454713
Hs.514107
Hs.512683
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID
s198812
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCL3L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001437.3
NP_001001437.2
3
696
GenBank mRNA
BC007783.2
-
3
691
4308472
Gene Symbol: CCL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002983.2
NP_002974.1
3
719
GenBank mRNA
BC071834.1
-
3
691
5935287
BX103455.1
-
3
719
D00044.1
-
3
700
M23452.1
-
3
700
M25315.1
-
3
700
X03754.1
-
3
700
Gene Symbol: CCL3L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021006.5
NP_066286.1
3
706
NR_111964.1
3
898
GenBank mRNA
BC027888.1
-
3
694
5222247
BC107710.1
-
3
710
6699133
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
monocyte chemotaxis
inflammatory response
G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
neutrophil chemotaxis
positive regulation of GTPase activity
lymphocyte chemotaxis
positive regulation of inflammatory response
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
MAPK cascade
osteoblast differentiation
cell activation
calcium ion transport
cellular calcium ion homeostasis
exocytosis
chemotaxis
cytoskeleton organization
cell-cell signaling
behavior
regulation of cell shape
response to toxic substance
positive regulation of gene expression
negative regulation of gene expression
T cell chemotaxis
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
calcium-mediated signaling
signaling
positive regulation of cell migration
negative regulation of bone mineralization
lipopolysaccharide-mediated signaling pathway
positive regulation of tumor necrosis factor production
eosinophil degranulation
protein kinase B signaling
positive regulation of neuron apoptotic process
astrocyte cell migration
negative regulation by host of viral transcription
negative regulation of osteoclast differentiation
eosinophil chemotaxis
macrophage chemotaxis
positive regulation of interleukin-1 beta secretion
positive regulation of calcium-mediated signaling
positive chemotaxis
positive regulation of protein kinase B signaling
positive regulation of calcium ion transport
regulation of sensory perception of pain
response to cholesterol
cellular response to organic cyclic compound
granulocyte chemotaxis
positive regulation of calcium ion import
positive regulation of natural killer cell chemotaxis
chemokine activity
CCR chemokine receptor binding
protein kinase activity
calcium-dependent protein kinase C activity
protein binding
phospholipase activator activity
kinase activity
CCR1 chemokine receptor binding
CCR5 chemokine receptor binding
chemoattractant activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System