Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERN1 SIRNA Assays ›
          Gene Symbol:
          ERN1
          Gene Name:
          endoplasmic reticulum to nucleus signaling 1
          Gene Aliases:
          IRE1, IRE1P, IRE1a, hIRE1p
          Chromosome Location:
          Chr.17: 64039142 - 64130819 on Build GRCh38
          UniGene ID:
          Hs.133982
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s200430
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001433.3 NP_001424.3 4, 5 403
          XM_017024347.1 XP_016879836.1 4, 5 1757
          XM_017024348.1 XP_016879837.1 4, 5 1757
          GenBank mRNA AB209869.1 - 4, 5 386
          AF059198.1 - 4, 5 386
          AK292403.1 - 4, 5 429
          BC130405.1 - 4, 5 336 40146436
          BC130407.1 - 4, 5 336 40146438
          DB128514.1 - 4, 5 403

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00176385_m1

          Panther Classification:

          Molecular Function -

          endoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          endothelial cell proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA cleavage
          mRNA catabolic process
          protein phosphorylation
          activation of signaling protein activity involved in unfolded protein response
          cell cycle arrest
          activation of JUN kinase activity
          regulation of macroautophagy
          positive regulation of RNA splicing
          response to endoplasmic reticulum stress
          cellular response to vascular endothelial growth factor stimulus
          peptidyl-serine autophosphorylation
          IRE1-mediated unfolded protein response
          protein autophosphorylation
          mRNA splicing, via endonucleolytic cleavage and ligation
          mRNA endonucleolytic cleavage involved in unfolded protein response
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to glucose stimulus
          RNA phosphodiester bond hydrolysis, endonucleolytic
          positive regulation of endoplasmic reticulum unfolded protein response
          insulin metabolic process
          peptidyl-serine trans-autophosphorylation
          magnesium ion binding
          endoribonuclease activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          enzyme binding
          Hsp70 protein binding
          identical protein binding
          protein homodimerization activity
          ADP binding
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0