Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
Gene Name:
ankyrin repeat domain 36
Gene Aliases:
UNQ2430
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
UniGene ID:
Hs.541894
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s200613
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
1
370
XM_006712514.3
XP_006712577.1
1
680
XM_011511130.1
XP_011509432.1
1
680
XM_011511131.1
XP_011509433.1
1
680
XM_017004009.1
XP_016859498.1
1
680
XM_017004010.1
XP_016859499.1
1
680
XM_017004011.1
XP_016859500.1
1
680
XM_017004012.1
XP_016859501.1
1
680
XM_017004013.1
XP_016859502.1
1
680
XM_017004014.1
XP_016859503.1
1
680
XM_017004015.1
XP_016859504.1
1
680
XM_017004016.1
XP_016859505.1
1
680
XM_017004017.1
XP_016859506.1
1
680
XM_017004018.1
XP_016859507.1
1
680
XM_017004019.1
XP_016859508.1
1
680
XM_017004020.1
XP_016859509.1
1
680
XM_017004021.1
XP_016859510.1
1
680
XM_017004022.1
XP_016859511.1
1
680
XM_017004023.1
XP_016859512.1
1
680
XM_017004024.1
XP_016859513.1
1
680
XM_017004025.1
XP_016859514.1
1
680
XM_017004026.1
XP_016859515.1
1
680
XM_017004027.1
XP_016859516.1
1
680
XM_017004028.1
XP_016859517.1
1
680
XM_017004029.1
XP_016859518.1
1
680
XM_017004030.1
XP_016859519.1
1
680
XM_017004031.1
XP_016859520.1
1
680
XM_017004032.1
XP_016859521.1
1
680
XM_017004033.1
XP_016859522.1
1
680
XM_017004034.1
XP_016859523.1
1
680
XM_017004035.1
XP_016859524.1
1
680
XM_017004036.1
XP_016859525.1
1
680
XM_017004037.1
XP_016859526.1
1
680
XM_017004038.1
XP_016859527.1
1
680
XM_017004039.1
XP_016859528.1
1
680
XM_017004040.1
XP_016859529.1
1
680
XM_017004041.1
XP_016859530.1
1
680
XM_017004042.1
XP_016859531.1
1
680
XM_017004043.1
XP_016859532.1
1
680
XM_017004044.1
XP_016859533.1
1
680
XM_017004045.1
XP_016859534.1
1
680
XM_017004046.1
XP_016859535.1
1
680
XM_017004047.1
XP_016859536.1
1
680
XM_017004048.1
XP_016859537.1
1
680
XM_017004049.1
XP_016859538.1
1
680
GenBank mRNA
AK304740.1
-
1
310
AK307629.1
-
1
277
AK316541.1
-
1
310
AY358571.1
-
1
369
BC046186.1
-
1
249
BC128262.1
-
1
209
BC128517.1
-
1
209
40118468
BX647769.1
-
1
315
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-swfkw:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0