Search
Search

Camk2a| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286809.1 | NP_001273738.1 | 4 | 605 | ||
| NM_009792.3 | NP_033922.1 | 5 | 638 | |||
| NM_177407.4 | NP_803126.1 | 16 | 1326 | |||
| GenBank mRNA | AK083245.1 | - | 16 | 1327 | ||
| AK122410.2 | - | 16 | 1322 | |||
| BB659027.1 | - | 4 | 561 | |||
| BC031745.1 | - | 16 | 1307 | 4506005 | ||
| X14836.1 | - | 16 | 1310 | |||
| X87142.1 | - | 5 | 638 | |||