Search Thermo Fisher Scientific
Gene Symbol: Epha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007938.2 | NP_031964.2 | 3 | 795 | ||
GenBank mRNA | AK090366.1 | - | 3 | 796 | ||
BC141090.1 | - | 3 | 311 | 9055702 | ||
BC145362.1 | - | 4 | 431 | |||
U58332.1 | - | 3 | 511 |