Search Thermo Fisher Scientific
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008076.3 | NP_032102.2 | 1 | 120 | ||
GenBank mRNA | AF024621.1 | - | 1 | 233 | ||
BC057957.1 | - | 1 | 105 | 5357713 | ||
CK618173.1 | - | 1 | 120 |