Search
Search

Daam1| Gene Symbol: Daam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286452.1 | NP_001273381.1 | 14, 15 | 2204 | ||
| NM_026102.3 | NP_080378.2 | 14, 15 | 2172 | |||
| NM_172464.3 | NP_766052.2 | 14, 15 | 2037 | |||
| GenBank mRNA | AK053785.1 | - | 14, 15 | 2093 | ||
| AK129185.1 | - | 14, 15 | 2015 | |||
| AK147480.1 | - | 14, 15 | 2018 | |||
| AY426535.1 | - | 14, 15 | 2082 | |||
| BC025638.1 | - | 13, 14 | 1934 | |||
| BC032287.1 | - | 1, 2 | 108 | |||
| BC038403.1 | - | 14, 15 | 1978 | |||
| BC047431.1 | - | 14, 15 | 1959 | |||
| BC076585.1 | - | 14, 15 | 2027 | 30622306 | ||