Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 23 | 3228 | ||
XM_017024011.1 | XP_016879500.1 | 23 | 3193 | |||
XM_017024012.1 | XP_016879501.1 | 23 | 3297 | |||
XM_017024013.1 | XP_016879502.1 | 23 | 3117 | |||
XM_017024014.1 | XP_016879503.1 | 23 | 3717 | |||
XM_017024015.1 | XP_016879504.1 | 20 | 2740 | |||
GenBank mRNA | AF423307.1 | - | 22 | 3085 | ||
AY028899.1 | - | 23 | 3228 | |||
CR627382.1 | - | 22 | 3034 |