Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BTN2A2 SIRNA Assays ›
Gene Symbol:
BTN2A2
Gene Name:
butyrophilin subfamily 2 member A2
Gene Aliases:
BT2.2, BTF2, BTN2.2
Chromosome Location:
Chr.6: 26382891 - 26394874 on Build GRCh38
UniGene ID:
Hs.373938
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20308
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BTN2A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197237.1
NP_001184166.1
5, 6
1070
NM_001197238.1
NP_001184167.1
5, 6
1171
NM_001197239.1
NP_001184168.1
3, 4
418
NM_001197240.1
NP_001184169.1
4, 5
766
NM_006995.4
NP_008926.2
5, 6
1048
NM_181531.2
NP_853509.1
4, 5
700
XM_005248797.3
XP_005248854.1
5, 6
1065
XM_005248798.4
XP_005248855.1
4, 5
1025
XM_006714953.3
XP_006715016.1
5, 6
1317
XM_006714955.3
XP_006715018.1
5, 6
1065
XM_011514228.2
XP_011512530.1
5, 6
1370
XM_011514229.2
XP_011512531.1
5, 6
879
XM_011514231.2
XP_011512533.1
3, 4
743
XM_017010166.1
XP_016865655.1
5, 6
1068
XM_017010167.1
XP_016865656.1
4, 5
970
XM_017010168.1
XP_016865657.1
5, 6
1321
XM_017010169.1
XP_016865658.1
5, 6
1039
GenBank mRNA
AK123902.1
-
1, 2
115
AK289883.1
-
4, 5
688
AK293526.1
-
3, 4
393
AK298570.1
-
4, 5
741
BC013258.2
-
3, 4
620
BC014021.2
-
5, 6
1046
4124138
BC017497.1
-
5, 6
1063
4648935
BI754184.1
-
3, 4
394
U90550.1
-
5, 6
1023
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cellular metabolic process
negative regulation of activated T cell proliferation
negative regulation of cytokine secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline