Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Xlr5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031493.1 | NP_113681.1 | 4, 5 | 340 | ||
GenBank mRNA | BC119509.2 | - | 4, 5 | 340 | 40053468 |
Gene Symbol: Xlr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045539.2 | NP_001039004.2 | 5, 6 | 472 | ||
GenBank mRNA | BC051681.1 | - | 5, 6 | 472 | 30021721 |
Gene Symbol: Xlr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111293.1 | NP_001104763.1 | 5, 6 | 427 |