Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mslnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177822.3 | NP_808490.1 | 4 | 585 | ||
GenBank mRNA | AK028891.1 | - | 4 | 585 | ||
BC052484.1 | - | 3 | 321 | 6312790 |