Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001186.3 | NP_001001186.3 | 4 | 707 | ||
GenBank mRNA | AK164227.1 | - | 4 | 708 | ||
BC141197.1 | - | 4 | 690 | 9055809 |