Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPSA SIRNA Assays ›
          Gene Symbol:
          Rpsa
          Gene Name:
          ribosomal protein SA
          Gene Aliases:
          67kDa, 67lr, AL022858, Lamr, Lamr1, Lamrl1, MLR, P40, P40-3, P40-8
          Chromosome Location:
          Chr.9: 120127766 - 120132369 on Build GRCm38
          UniGene ID:
          Mm.311972
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s203656

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpsa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011029.4 NP_035159.3 3 306
          GenBank mRNA AF140348.1 - 3 293
          AK010423.1 - 3 286
          AK010985.1 - 3 288
          AK011041.1 - 3 288
          AK011043.1 - 3 288
          AK075778.1 - 1 18
          AK075790.1 - 1 18
          AK082935.1 - 3 285
          AK088954.1 - 3 286
          AK134224.1 - 3 286
          AK135488.1 - 3 288
          AK145713.1 - 3 283
          AK151123.1 - 3 285
          AK160551.1 - 3 285
          AK160625.1 - 3 285
          AK165219.1 - 3 286
          AK166697.1 - 3 285
          AK166874.1 - 3 288
          AK167132.1 - 3 286
          AK205896.1 - 2 191
          AK212093.1 - 2 191
          AK212679.1 - 2 191
          AK213565.1 - 2 191
          AK214711.1 - 2 191
          AK215661.1 - 2 191
          AK216125.1 - 2 191
          AK218136.1 - 2 191
          AK219743.1 - 2 191
          AK219820.1 - 2 191
          AV586849.1 - 3 383
          BC003829.1 - 3 270 3600738
          BC037195.1 - 2 233 4500748
          BC055886.1 - 2 440 4240083
          BC081461.1 - 2 241 6820702
          BC084677.1 - 3 270 6476344
          BC092041.1 - 3 271 2651126
          BC110285.1 - 3 271 5151374
          EU554633.1 - 2 208
          J02870.1 - 3 287
          X06406.1 - 3 285

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          rRNA export from nucleus
          translation
          single organismal cell-cell adhesion
          structural constituent of ribosome
          laminin receptor activity
          ribosome binding
          laminin binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-w4hmg:80/100.66.74.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0