Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PYCARD SIRNA Assays ›
          Gene Symbol:
          Pycard
          Gene Name:
          PYD and CARD domain containing
          Gene Aliases:
          9130417A21Rik, Asc, CARD5, TMS-1, TNS1, masc
          Chromosome Location:
          Chr.7: 127991373 - 127993867 on Build GRCm38
          UniGene ID:
          Mm.24163
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s205578

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pycard
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_023258.4 NP_075747.3 1 469
          GenBank mRNA AB032249.1 - 1 341
          AF310104.1 - 1 281
          AK007742.1 - 1 339
          AK009852.1 - 1 344
          AK018682.1 - 1 349
          AK152677.1 - 1 329
          BC008252.1 - 1 311 2648391

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          myeloid dendritic cell activation
          activation of innate immune response
          positive regulation of defense response to virus by host
          myeloid dendritic cell activation involved in immune response
          immune system process
          positive regulation of antigen processing and presentation of peptide antigen via MHC class II
          positive regulation of adaptive immune response
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          response to bacterium
          regulation of autophagy
          regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of actin filament polymerization
          regulation of protein stability
          negative regulation of NF-kappaB transcription factor activity
          interleukin-1 beta production
          negative regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          tumor necrosis factor-mediated signaling pathway
          positive regulation of activated T cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of GTPase activity
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          macropinocytosis
          innate immune response
          positive regulation of JNK cascade
          positive regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          positive regulation of phagocytosis
          defense response to Gram-negative bacterium
          positive regulation of T cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          negative regulation of protein serine/threonine kinase activity
          intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of chemokine secretion
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of T cell migration
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          positive regulation of cysteine-type endopeptidase activity
          positive regulation of interleukin-10 secretion
          positive regulation of extrinsic apoptotic signaling pathway
          regulation of intrinsic apoptotic signaling pathway
          protease binding
          interleukin-6 receptor binding
          protein binding
          tropomyosin binding
          peptidase activator activity involved in apoptotic process
          myosin I binding
          enzyme binding
          Pyrin domain binding
          protein homodimerization activity
          ion channel binding
          protein dimerization activity
          BMP receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0