Search
Search

Rps20| Gene Symbol: Rps20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026147.5 | NP_080423.1 | 4 | 305 | ||
| GenBank mRNA | AK002405.1 | - | 4 | 306 | ||
| AK014594.1 | - | 305 | ||||
| AK136398.1 | - | 4 | 308 | |||
| AK145656.1 | - | 4 | 306 | |||
| AK150867.1 | - | 4 | 305 | |||
| AK150972.1 | - | 4 | 305 | |||
| BC011323.1 | - | 4 | 292 | 3257215 | ||
| BC090389.1 | - | 4 | 296 | 3482252 | ||