Search
Search

Dnajc6| Gene Symbol: Dnajc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164583.1 | NP_001158055.1 | 16 | 2537 | ||
| NM_001164584.1 | NP_001158056.1 | 16 | 2482 | |||
| NM_001164585.1 | NP_001158057.1 | 17 | 2547 | |||
| NM_198412.2 | NP_940804.1 | 17 | 2456 | |||
| GenBank mRNA | AK012831.1 | - | 7 | 1201 | ||
| AK049935.1 | - | 4 | 499 | |||
| AK122293.1 | - | 16 | 2458 | |||
| AK147570.1 | - | 17 | 2419 | |||
| AK147637.1 | - | 17 | 2419 | |||
| AK163657.1 | - | 16 | 2356 | |||
| BC035262.1 | - | 11 | 1691 | |||
| BC060734.1 | - | 16 | 2537 | 6409942 | ||
| BC064460.1 | - | 17 | 2425 | 6821822 | ||