Search
Search

Hepacam| Gene Symbol: Hepacam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175189.4 | NP_780398.2 | 4 | 792 | ||
| GenBank mRNA | AK013637.1 | - | 4 | 792 | ||
| AK081154.1 | - | 4 | 708 | |||
| AK138438.1 | - | 3 | 316 | |||
| AK158265.1 | - | 4 | 1030 | |||
| BC082537.1 | - | 4 | 706 | |||
| BC139057.1 | - | 4 | 782 | 8862079 | ||
| BC139058.1 | - | 4 | 782 | 8862080 | ||