Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20770
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
22
2231
NM_001034955.1
NP_001030127.1
22
2297
NM_001034956.1
NP_001030128.1
19
1784
NM_001034957.1
NP_001030129.1
14
1349
NM_001290294.1
NP_001277223.1
22
2280
NM_001290295.1
NP_001277224.1
17
1563
NM_001290296.1
NP_001277225.1
20
2141
NM_001290297.1
NP_001277226.1
17
1562
NM_001290298.1
NP_001277227.1
18
1667
NM_006434.2
NP_006425.2
14
1349
NM_015385.3
NP_056200.1
18
1932
NM_024991.2
NP_079267.1
18
1659
XM_006717589.1
XP_006717652.1
22
2281
XM_006717593.1
XP_006717656.1
16
1514
XM_011539140.1
XP_011537442.1
24
3025
XM_011539150.1
XP_011537452.1
23
2950
XM_011539155.1
XP_011537457.1
18
2600
XM_011539167.1
XP_011537469.1
17
1883
XM_017015500.1
XP_016870989.1
20
2698
XM_017015501.1
XP_016870990.1
23
2950
XM_017015502.1
XP_016870991.1
22
2857
XM_017015503.1
XP_016870992.1
21
2782
XM_017015504.1
XP_016870993.1
22
1939
XM_017015505.1
XP_016870994.1
20
2698
XM_017015506.1
XP_016870995.1
21
1864
XM_017015507.1
XP_016870996.1
22
2857
XM_017015508.1
XP_016870997.1
20
1771
XM_017015509.1
XP_016870998.1
20
1793
XM_017015510.1
XP_016870999.1
20
2698
XM_017015511.1
XP_016871000.1
19
1687
XM_017015512.1
XP_016871001.1
17
1598
XM_017015513.1
XP_016871002.1
17
1589
XM_017015514.1
XP_016871003.1
18
1673
XM_017015515.1
XP_016871004.1
21
2140
XM_017015516.1
XP_016871005.1
20
2113
XM_017015517.1
XP_016871006.1
20
2056
XM_017015518.1
XP_016871007.1
20
2069
XM_017015519.1
XP_016871008.1
19
2042
XM_017015520.1
XP_016871009.1
19
1981
XM_017015521.1
XP_016871010.1
19
1994
XM_017015522.1
XP_016871011.1
18
1954
XM_017015523.1
XP_016871012.1
18
1910
XM_017015524.1
XP_016871013.1
17
1883
XM_017015525.1
XP_016871014.1
21
1868
XM_017015526.1
XP_016871015.1
20
1771
XM_017015527.1
XP_016871016.1
20
1793
XM_017015528.1
XP_016871017.1
19
1744
XM_017015529.1
XP_016871018.1
19
1696
XM_017015530.1
XP_016871019.1
19
1687
XM_017015531.1
XP_016871020.1
19
1700
XM_017015532.1
XP_016871021.1
18
1669
XM_017015533.1
XP_016871022.1
18
1673
XM_017015534.1
XP_016871023.1
20
1771
XM_017015535.1
XP_016871024.1
18
1625
XM_017015536.1
XP_016871025.1
18
1616
XM_017015537.1
XP_016871026.1
17
1585
XM_017015538.1
XP_016871027.1
19
1744
XM_017015539.1
XP_016871028.1
17
1598
XM_017015540.1
XP_016871029.1
17
1589
GenBank mRNA
AB037717.1
-
18
1591
AF136380.1
-
13
1304
AF136381.1
-
19
2096
AF330623.1
-
16
1517
AF330624.1
-
2
215
AF356525.1
-
22
2242
AF356526.1
-
22
2308
AF356527.1
-
19
1795
AJ489942.1
-
17
1622
AK294924.1
-
6
541
AK300407.1
-
17
1516
AK304130.1
-
6
604
AK304753.1
-
18
2575
AL117472.1
-
18
1890
AM260536.1
-
22
2238
BC042612.1
-
16
1956
BC152463.1
-
18
1591
8862599
BX640814.1
-
6
1594
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-68wbn:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0