Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RFPL2 SIRNA Assays ›
          Gene Symbol:
          RFPL2 RFPL1 RFPL3
          Gene Name:
          ret finger protein like 2
          ret finger protein like 1
          ret finger protein like 3
          Gene Aliases:
          RNF79
          RNF78
          -
          Chromosome Location:
          Chr.22: 32190435 - 32204844 on Build GRCh38
          Chromosome Location:
          Chr.22: 29403031 - 29442491 on Build GRCh38
          Chromosome Location:
          Chr.22: 32354885 - 32361161 on Build GRCh38
          UniGene ID:
          Hs.157427
          Hs.648249
          Hs.558455
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s21097

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RFPL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098527.2 NP_001091997.2 5 1598
          NM_001159545.1 NP_001153017.1 4 640
          NM_001159546.1 NP_001153018.1 4 669
          NM_006605.3 NP_006596.2 2 684
          XM_011529826.1 XP_011528128.1 5 1802
          XM_011529827.2 XP_011528129.1 6 977
          XM_011529828.1 XP_011528130.1 6 1712
          XM_011529830.2 XP_011528132.1 6 1062
          XM_011529832.1 XP_011528134.1 4 1688
          XM_017028535.1 XP_016884024.1 5 807
          XM_017028536.1 XP_016884025.1 5 856
          GenBank mRNA AJ010231.1 - 2 684
          AK055999.1 - 5 1598
          BC051910.1 - 4 640 5729832
          BC069737.1 - 2 606 7262638
          BC069827.1 - 2 605 7262662
          CR456563.1 - 4 630
          Gene Symbol: RFPL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021026.2 NP_066306.2 2 688
          XM_011530298.2 XP_011528600.1 4 648
          XM_011530299.2 XP_011528601.1 3 550
          XM_017028900.1 XP_016884389.1 6 1035
          XM_017028901.1 XP_016884390.1 5 969
          XM_017028902.1 XP_016884391.1 10 4013
          GenBank mRNA AJ010228.1 - 2 688
          AJ010229.1 - 2 685
          BC104768.1 - 2 415
          CR456562.2 - 2 507
          Gene Symbol: RFPL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098535.1 NP_001092005.1 2 684
          NM_006604.2 NP_006595.1 2 410
          GenBank mRNA AJ010232.1 - 2 684
          AK097738.1 - 3 1162
          BC031689.1 - 2 410 5167710
          BC069753.1 - 2 608 7262650
          CR456564.1 - 2 526

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          metal ion binding
          zinc ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0