Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PHTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323041.1 | NP_001309970.1 | 18 | 2856 | ||
NM_001323042.1 | NP_001309971.1 | 18 | 2340 | |||
NM_001323043.1 | NP_001309972.1 | 18 | 2741 | |||
NM_001323044.1 | NP_001309973.1 | 18 | 3114 | |||
NM_001323045.1 | NP_001309974.1 | 19 | 2825 | |||
NM_001323046.1 | NP_001309975.1 | 15 | 2352 | |||
NM_006608.2 | NP_006599.2 | 17 | 2645 | |||
NR_136564.1 | 20 | 2991 | ||||
NR_136565.1 | 18 | 2632 | ||||
NR_136566.1 | 19 | 2907 | ||||
NR_136567.1 | 19 | 2792 | ||||
GenBank mRNA | AJ011863.1 | - | 17 | 2224 | ||
AK126773.1 | - | 11 | 3344 | |||
DA728266.1 | - | 6 | 568 |