Search Thermo Fisher Scientific
Gene Symbol: ZNF275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080485.3 | NP_001073954.3 | 4 | 829 | ||
GenBank mRNA | AK122976.1 | - | 4 | 829 | ||
AK127133.1 | - | 1 | 527 | |||
BC001545.1 | - | 3 | 1474 | |||
BC136634.1 | - | 2 | 1855 | 9020624 |